Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 500 mm
50 x 100 mm
Catalog number 000097.GD
also known as 000097-GD
Price (USD)/ Each
581.65
Online exclusive
646.00 
Save 64.35 (10%)
-
Add to cart
Quantity:
50 x 500 mm
Request bulk or custom format
Price (USD)/ Each
581.65
Online exclusive
646.00 
Save 64.35 (10%)
Add to cart
Copper foil, 0.25mm (0.01in) thick, hard, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Catalog number000097.GD
Price (USD)/ Each
581.65
Online exclusive
646.00 
Save 64.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.25 mm (0.01 in.) thick
Total Metal Impurities≤0.001%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only