Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals
Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 000224.A1
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-100 mesh (less than 150 micron)
Trace Metal99+% (metals basa excluding Co) Co 0.1% max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only