Platinum foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx.0.34g/25 x 25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 000262.GH
also known as 000262-GH
Price (USD)/ Each
1,525.65
Online exclusive
1695.00 
Save 169.35 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
1,525.65
Online exclusive
1695.00 
Save 169.35 (10%)
Add to cart
Platinum foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number000262.GH
Price (USD)/ Each
1,525.65
Online exclusive
1695.00 
Save 169.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metalsless than 0.1% trace metals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only