Thermo Scientific Chemicals

Tantalum foil, 0.025mm (0.001in) thick, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 000335.FF
25 x 25 mm, Each
Thermo Scientific Chemicals

Tantalum foil, 0.025mm (0.001in) thick, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 000335.FF
25 x 25 mm, Each
Quantity
Catalog number: 000335.FF
also known as 000335-FF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-25-7
IUPAC Name
tantalum
Molecular Formula
Ta
InChI Key
GUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES
[Ta]
Appearance (Color)
Silvery
Form
Foil, 0.025mm (0.001in) thick.
Assay from Supplier's CofA
99.997% (metals basis)
Assay from Supplier's CofA
≥99.997% (metals basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text