Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

packaged under argon in glass bottle
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 000408.09
also known as 000408-09
Price (USD)/ Each
449.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
449.00
Add to cart
Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Catalog number000408.09
Price (USD)/ Each
449.00
-
Add to cart
Chemical Identifiers
CAS7440-64-4
IUPAC Nameytterbium
Molecular FormulaYb
InChI KeyNAWDYIZEMPQZHO-UHFFFAOYSA-N
SMILES[Yb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only