Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 000428.14
also known as 000428-14
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
Add to cart
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number000428.14
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.0001%
FormShot, 1.5-5 mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only