Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 000472.36
also known as 000472-36
Price (USD)/ Each
285.65
Online exclusive
336.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
285.65
Online exclusive
336.00 
Save 50.35 (15%)
Add to cart
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis), Thermo Scientific Chemicals
Catalog number000472.36
Price (USD)/ Each
285.65
Online exclusive
336.00 
Save 50.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
Molecular Weight (g/mol)52.00
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces, 3-12mm
Total Metal Impurities≤0.8%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only