Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO), Thermo Scientific Chemicals
Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO), Thermo Scientific Chemicals

Form: Approx. 1.8g/25 x 25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 000616.FI
also known as 000616-FI
Price (USD)/ Each
627.65
Online exclusive
697.00 
Save 69.35 (10%)
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
627.65
Online exclusive
697.00 
Save 69.35 (10%)
Add to cart
Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO), Thermo Scientific Chemicals
Catalog number000616.FI
Price (USD)/ Each
627.65
Online exclusive
697.00 
Save 69.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-65-5
IUPAC Nameyttrium
Molecular FormulaY
InChI KeyVWQVUPCCIRVNHF-UHFFFAOYSA-N
SMILES[Y]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only