Aluminum shot, irregular, 15mm and down, 99.9% (metals basis), Thermo Scientific Chemicals
Aluminum shot, irregular, 15mm and down, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum shot, irregular, 15mm and down, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number 000632.A3
also known as 000632-A3
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
Aluminum shot, irregular, 15mm and down, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number000632.A3
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Particle Size15mm and down
Trace Metalsless than 0.1% trace metals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only