Zinc ingot, 345mm (13.6in) long, 99.99% (metals basis), Thermo Scientific Chemicals
Zinc ingot, 345mm (13.6in) long, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc ingot, 345mm (13.6in) long, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Pc.
1 Pc.
Catalog number 000682.KJ
also known as 000682-KJ
Price (USD)/ Each
149.00
-
Add to cart
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)/ Each
149.00
Add to cart
Zinc ingot, 345mm (13.6in) long, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number000682.KJ
Price (USD)/ Each
149.00
-
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities< 0.01%
FormMetallic bar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only