Platinum shot, 2.4mm (0.09in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum shot, 2.4mm (0.09in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum shot, 2.4mm (0.09in) & down, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 000719.06
also known as 000719-06
Price (USD)/ Each
1,007.65
Online exclusive
1119.00 
Save 111.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
1,007.65
Online exclusive
1119.00 
Save 111.35 (10%)
Add to cart
Platinum shot, 2.4mm (0.09in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number000719.06
Price (USD)/ Each
1,007.65
Online exclusive
1119.00 
Save 111.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver to gray
Total Metal Impurities0.1% max.
Particle Size2.4mm and down
FormShot
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only