Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 000908.36
also known as 000908-36
Price (USD)/ Each
52.65
Online exclusive
58.40 
Save 5.75 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
58.40 
Save 5.75 (10%)
Add to cart
Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number000908.36
Price (USD)/ Each
52.65
Online exclusive
58.40 
Save 5.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-40+100 mesh (150 - 425 micron)
Trace Metalsless than 0.5% trace metals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only