Tin powder, -100 mesh, 99.85% (metals basis), Thermo Scientific Chemicals
Tin powder, -100 mesh, 99.85% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin powder, -100 mesh, 99.85% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 000941.36
also known as 000941-36
Price (USD)/ Each
162.65
Online exclusive
181.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
162.65
Online exclusive
181.00 
Save 18.35 (10%)
Add to cart
Tin powder, -100 mesh, 99.85% (metals basis), Thermo Scientific Chemicals
Catalog number000941.36
Price (USD)/ Each
162.65
Online exclusive
181.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-100 mesh (less than 150 micron)
Trace Metal0.15%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only