DRI™ Cocaine Drugs of Abuse Assays
DRI™ Cocaine Drugs of Abuse Assays
Actual product may vary
Thermo Scientific™

DRI™ Cocaine Drugs of Abuse Assays

Save time in reagent preparation with liquid, ready-to-use assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00551 x 100 mL
00561 x 500 mL
100145933 x 18 mL
3 Options
Catalog number 0055
Price (USD)/ Each
-
Request A Quote
Quantity:
1 x 100 mL
Save time in reagent preparation with liquid, ready-to-use assays. Thermo Scientific™ DRI™ Cocaine Drugs of Abuse Assays offer applications for an array of clinical chemistry analyzers. These assays have been field-tested on a wide range of analyzers to deliver high analytical accuracy and are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Cocaine Drugs of Abuse Assays
Detectable AnalytesBenzoylecgonine
DoA CalibratorsDRI Multi-drug Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 100 mL
Unit SizeEach
Contents & Storage
2°C to 8°C