Filter for GeneTitan™ fluidics bottles
Product Image
Applied Biosystems™
Filter for GeneTitan™ fluidics bottles
This filter is used with the GeneTitan™ HT Microarray System. Each GeneTitan MC Instrument requires three filters.The GeneTitan HT MicroarrayRead more
Have Questions?
Catalog number 01-0671
Price (USD)/ 1 each
34.08
-
Add to cart
Price (USD)/ 1 each
34.08
Add to cart
Filter for GeneTitan™ fluidics bottles
Catalog number01-0671
Price (USD)/ 1 each
34.08
-
Add to cart
This filter is used with the GeneTitan™ HT Microarray System. Each GeneTitan MC Instrument requires three filters.
The GeneTitan HT Microarray System automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument.
The imaging device in the GeneTitan HT Microarray System uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in genome-wide expression and genotyping studies.
The GeneTitan HT Microarray System provides the highest throughput and laboratory productivity per technician by minimizing user intervention and allowing unattended, overnight processing of large numbers of samples in parallel. This unique degree of hands-free automation means your data will be consistently reproducible, your lab will be more productive, and you can spend less time acquiring and managing data and have more time for science.
Key benefits
Easy – Designed for operators with minimal microarray experience
Flexible – Supports gene expression on 16-, 24-, 96-array layouts and genotyping studies on 96- and 384-array layouts
Accurate – Delivers high-quality, consistent data every time
Efficient – Condenses hands-on processing time to as little as 30 minutes, images an array in less than 5 minutes, and operates unattended overnight
Robust – Has fewer moving parts so it's easily maintained
Scalable – Meets both medium- and high-throughput needs
Adaptable – Creates flexible workflows and sample registration via Affymetrix GeneChip Command Console Software (AGCC)
To learn more about how the GeneTitan HT Microarray System can maximize productivity and data reproducibility in your lab, please contact your local distributor for pricing and information.
Related Links
GeneTitan™ ZeroStat AntiStatic Gun
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneTitan Multi-Channel (MC) Instrument
For Use With (Application)GeneTitan Multi-Channel (MC) Instrument Accessory
Unit Size1 each