Thermo Scientific Chemicals

Cesium chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010018.14
25 g, Each
Thermo Scientific Chemicals

Cesium chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010018.14
25 g, Each
Quantity
Catalog number: 010018.14
also known as 010018-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-17-8
IUPAC Name
caesium(1+) chloride
Molecular Formula
ClCs
InChI Key
AIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES
[Cl-].[Cs+]
Total Metal Impurities
0.1% max.

Description

Analytical chemistry reagent for detecting inorganic ions. Source of cesium metal. Phase transfer catalyst in organic chemistry. Used for density gradient work to separate and purify proteins (isopycnic centrifugation) including chromosomal, plasmid and organelle (mitochondrial or plasmid) DNAs, and different RNA types. Preparation of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text