Thermo Scientific Chemicals

Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010019.88
2.5 g, Each
Thermo Scientific Chemicals

Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010019.88
2.5 g, Each
Quantity
Catalog number: 010019.88
also known as 010019-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13400-13-0
IUPAC Name
caesium(1+) fluoride
Molecular Formula
CsF
InChI Key
XJHCXCQVJFPJIK-UHFFFAOYSA-M
SMILES
[F-].[Cs+]
Total Metal Impurities
0.1% max.

Description

Used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text