Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals
Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010022.14
Price (USD)/ Each
78.65
Online exclusive
86.80 
Save 8.15 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
86.80 
Save 8.15 (9%)
Add to cart
Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number010022.14
Price (USD)/ Each
78.65
Online exclusive
86.80 
Save 8.15 (9%)
-
Add to cart
Chemical Identifiers
CAS7789-17-5
IUPAC Namecaesium(1+) iodide
Molecular FormulaCsI
InChI KeyXQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES[I-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cesium iodide is used in FT-IR (Fourier Transform Infrared Spectrometers) as a beamsplitter. Cesium Iodide exhibits high gamma-ray stopping power. Useful as input phosphor in fluoroscopy equipment, and efficient photocathode for extreme ultraviolet wavelengths. Fast and dense scintillating material - useful in electromagnetic calorimetry in particle physics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only