Thermo Scientific Chemicals

Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010022.22
100 g, Each
Thermo Scientific Chemicals

Cesium iodide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 010022.22
100 g, Each
Quantity
Catalog number: 010022.22
also known as 010022-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-17-5
IUPAC Name
caesium(1+) iodide
Molecular Formula
CsI
InChI Key
XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES
[I-].[Cs+]
Total Metal Impurities
0.1% max.

Description

Cesium iodide is used in FT-IR (Fourier Transform Infrared Spectrometers) as a beamsplitter. Cesium Iodide exhibits high gamma-ray stopping power. Useful as input phosphor in fluoroscopy equipment, and efficient photocathode for extreme ultraviolet wavelengths. Fast and dense scintillating material - useful in electromagnetic calorimetry in particle physics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text