Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
100 x 500 mm
Catalog number 010047.GH
also known as 010047-GH
Price (USD)/ Each
441.65
Online exclusive
490.00 
Save 48.35 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
441.65
Online exclusive
490.00 
Save 48.35 (10%)
Add to cart
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010047.GH
Price (USD)/ Each
441.65
Online exclusive
490.00 
Save 48.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only