Thermo Scientific Chemicals
Platinum Iridium wire, 0.5mm (0.02in) dia, hard, Thermo Scientific Chemicals
Catalog number: 010054.BW
50 cm, Each
Thermo Scientific Chemicals
Platinum Iridium wire, 0.5mm (0.02in) dia, hard, Thermo Scientific Chemicals
Catalog number: 010054.BW
50 cm, Each
Quantity
Catalog number: 010054.BW
also known as 010054-BW
Price (USD)
Quantity
-
Have Questions?
Description
Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.