Thermo Scientific Chemicals
Platinum 10 wt% Rhodium wire, 0.25mm (0.01in) dia, ISA Type S Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 010065.CC
500 cm, Each
Thermo Scientific Chemicals
Platinum 10 wt% Rhodium wire, 0.25mm (0.01in) dia, ISA Type S Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 010065.CC
500 cm, Each
Quantity
Catalog number: 010065.CC
also known as 010065-CC
Price (USD)
Have Questions?
Description
Platinum rhodium wire is used as a catalyst in preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.