Thermo Scientific Chemicals

Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals

Catalog number: 010103.22
100 g, Each
Thermo Scientific Chemicals

Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals

Catalog number: 010103.22
100 g, Each
Quantity
Catalog number: 010103.22
also known as 010103-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-39-3
IUPAC Name
barium
Molecular Formula
Ba
InChI Key
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
SMILES
[Ba]
Total Metal Impurities
1% max., Sr ≤0.8%

Description

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text