Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals
Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010130.A1
also known as 010130-A1
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
Add to cart
Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals
Catalog number010130.A1
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
-
Add to cart
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal75-82% C, 18-25% Ash
Particle Size-325 mesh (less than 45 micron)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only