Chromium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Chromium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number 010148.A3
also known as 010148-A3
Price (USD)/ Each
694.65
Online exclusive
772.00 
Save 77.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
694.65
Online exclusive
772.00 
Save 77.35 (10%)
Add to cart
Chromium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Catalog number010148.A3
Price (USD)/ Each
694.65
Online exclusive
772.00 
Save 77.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialChromium
ColorGray
CAS7440-47-3
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only