Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 010161.30
also known as 010161-30
Price (USD)/ Each
47.65
Online exclusive
53.30 
Save 5.65 (11%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.30 
Save 5.65 (11%)
Add to cart
Copper turnings, 99+% (metals basis), Thermo Scientific Chemicals
Catalog number010161.30
Price (USD)/ Each
47.65
Online exclusive
53.30 
Save 5.65 (11%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only