Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Quantity:
50 x 50 mm
Catalog number 010170.FI
also known as 010170-FI
Price (USD)/ Each
1,091.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
1,091.00
Add to cart
Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Catalog number010170.FI
Price (USD)/ Each
1,091.00
-
Add to cart
Chemical Identifiers
CAS7440-52-0
IUPAC Nameerbium
Molecular FormulaEr
InChI KeyUYAHIZSMUZPPFV-UHFFFAOYSA-N
SMILES[Er]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Erbium foil is commonly used as a photographic filter, and because of its resilience it is useful as a metallurgical additive. It is used as an alloying element with titanium and is also used to produce infra-red absorbing glass. Used in nuclear technology for control rods; an alloy in vanadium to lower hardness, lasers and erbium doped fiber amplifiers, a colorant in glass and porcelain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only