Tribenzylphosphine, Thermo Scientific Chemicals
Tribenzylphosphine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tribenzylphosphine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 010173.03
also known as 010173-03
Price (USD)/ Each
57.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
57.50
Add to cart
Tribenzylphosphine, Thermo Scientific Chemicals
Catalog number010173.03
Price (USD)/ Each
57.50
-
Add to cart
Specifications
Chemical Name or MaterialTribenzylphosphine
CAS7650-89-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Tribenzylphosphine is used as a co catalyst in preparation of coumarin derivatives via nickel-catalyzed cycloaddition, copolymerization reactions, Rh-catalyzed hydroformylation reactions, preparation of substituted allylamines by three-component coupling of alkynes, imines and organoboranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only