Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals
Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 010175.FF
also known as 010175-FF
Price (USD)/ Each
860.00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
860.00
Add to cart
Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals
Catalog number010175.FF
Price (USD)/ Each
860.00
-
Add to cart
Chemical Identifiers
CAS7440-53-1
IUPAC Nameeuropium
Molecular FormulaEu
InChI KeyOGPBJKLSAFTDLK-UHFFFAOYSA-N
SMILES[Eu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only