Tri-tert-butylphosphine, 98%, Thermo Scientific Chemicals
Tri-tert-butylphosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-tert-butylphosphine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 010178.06
also known as 010178-06
Price (USD)/ Each
342.65
Online exclusive
380.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
342.65
Online exclusive
380.00 
Save 37.35 (10%)
Add to cart
Tri-tert-butylphosphine, 98%, Thermo Scientific Chemicals
Catalog number010178.06
Price (USD)/ Each
342.65
Online exclusive
380.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13716-12-6
IUPAC Nametri-tert-butylphosphane
Molecular FormulaC12H27P
InChI KeyBWHDROKFUHTORW-UHFFFAOYSA-N
SMILESCC(C)(C)P(C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or clear colorless
Phosphorus-31 NMR>95.0%
FormFused solid or liquid as melt
Assay from Supplier's CofA>95.0% (by vendor's 31P-NMR)
Tri-tert-butylphosphine is used primarily in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only