This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010202.09
also known as 010202-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHafnium powder
ColorGray
CAS7440-58-6
Health Hazard 1H250
Health Hazard 2GHS H Statement H251 Self-heating: may catch fire.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.