Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
500 cm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010205.BY
also known as 010205-BY
Price (USD)/ Each
-
Quantity:
100 cm
Request bulk or custom format
Chemical Identifiers
CAS7440-58-6
IUPAC Namehafnium
Molecular FormulaHf
InChI KeyVBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES[Hf]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.05% excluding Zr, Zr nominal 3%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only