Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron powder, spherical, APS 6-10 micron, reduced, 99.5%, Thermo Scientific Chemicals

Apparent density 2.0-3.0g/cm3
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
500 g
2 x 500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010214.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)minimum 99.5%
Particle SizeAPS 6-10 micron
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only