Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals
Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific™

Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number 010236.A7
also known as 010236-A7
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
Add to cart
Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number010236.A7
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only