Nickel foil, 0.025mm (0.001in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Nickel foil, 0.025mm (0.001in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel foil, 0.025mm (0.001in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 250 mm
50 x 50 mm
Catalog number 010254.GB
also known as 010254-GB
Price (USD)/ Each
710.65
Online exclusive
789.00 
Save 78.35 (10%)
-
Add to cart
Quantity:
50 x 250 mm
Request bulk or custom format
Price (USD)/ Each
710.65
Online exclusive
789.00 
Save 78.35 (10%)
Add to cart
Nickel foil, 0.025mm (0.001in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number010254.GB
Price (USD)/ Each
710.65
Online exclusive
789.00 
Save 78.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialNickel foil
Name Note0.025mm (0.001 in.) thick
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only