Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Form: Approx.42.9g/50 x 100mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 x 500 mm
50 x 100 mm
100 x 200 mm
Catalog number 010257.HQ
also known as 010257-HQ
Price (USD)/ Each
1,288.00
-
Add to cart
Quantity:
200 x 500 mm
Request bulk or custom format
Price (USD)/ Each
1,288.00
Add to cart
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number010257.HQ
Price (USD)/ Each
1,288.00
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only