Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 200 mm
50 x 100 mm
200 x 500 mm
Catalog number 010259.GK
also known as 010259-GK
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
100 x 200 mm
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
Niobium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number010259.GK
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.8% (metals basis)
Appearance (Color)Silvery
FormFoil, 0.25mm (0.01in) thick, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only