Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Form: Approx.6.7g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
25 m
Catalog number 010264.G5
also known as 010264-G5
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number010264.G5
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only