Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010275.36
also known as 010275-36
Price (USD)/ Each
612.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
721.00 
Save 108.35 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
612.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
721.00 
Save 108.35 (15%)
Add to cart
Niobium powder, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number010275.36
Price (USD)/ Each
612.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
721.00 
Save 108.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Total Metal Impurities0.2% max.
Particle Size-325 mesh
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only