Niobium powder, -60 mesh, 99.8% (metals basis excluding Ta), Ta <1%, Thermo Scientific Chemicals
Niobium powder, -60 mesh, 99.8% (metals basis excluding Ta), Ta &lt;1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium powder, -60 mesh, 99.8% (metals basis excluding Ta), Ta <1%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010276.22
also known as 010276-22
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
Add to cart
Niobium powder, -60 mesh, 99.8% (metals basis excluding Ta), Ta <1%, Thermo Scientific Chemicals
Catalog number010276.22
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-60 mesh (less than 250 micron)
Trace MetalTotal metal impurities less than 0.2% excluding Ta, Ta <1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only