Palladium wire, 0.127mm (0.005in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Palladium wire, 0.127mm (0.005in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium wire, 0.127mm (0.005in) dia, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 m
2 m
Catalog number 010277.G6
also known as 010277-G6
Price (USD)/ Each
1,153.00
-
Add to cart
Quantity:
10 m
Request bulk or custom format
Price (USD)/ Each
1,153.00
Add to cart
Palladium wire, 0.127mm (0.005in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number010277.G6
Price (USD)/ Each
1,153.00
-
Add to cart
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.127mm (0.005in) diameter.
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Silver white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only