Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
100 x 100 mm
Catalog number 010282.FI
also known as 010282-FI
Price (USD)/ Each
999.65
Online exclusive
1111.00 
Save 111.35 (10%)
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
999.65
Online exclusive
1111.00 
Save 111.35 (10%)
Add to cart
Platinum gauze, 100 mesh woven from 0.0762mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number010282.FI
Price (USD)/ Each
999.65
Online exclusive
1111.00 
Save 111.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white guage
Recommended StorageAmbient temperatures
View more
It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only