Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 010283.FL
also known as 010283-FL
Price (USD)/ Each
1,326.65
Online exclusive
1474.00 
Save 147.35 (10%)
-
Add to cart
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)/ Each
1,326.65
Online exclusive
1474.00 
Save 147.35 (10%)
Add to cart
Platinum gauze, 52 mesh woven from 0.1mm (0.004in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number010283.FL
Price (USD)/ Each
1,326.65
Online exclusive
1474.00 
Save 147.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only