Platinum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Platinum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 cm
5 cm
Catalog number 010284.BO
also known as 010284-BO
Price (USD)/ Each
206.65
Online exclusive
229.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
1 cm
Request bulk or custom format
Price (USD)/ Each
206.65
Online exclusive
229.00 
Save 22.35 (10%)
Add to cart
Platinum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010284.BO
Price (USD)/ Each
206.65
Online exclusive
229.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only