Platinum wire, 0.2032mm (0.008in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Platinum wire, 0.2032mm (0.008in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum wire, 0.2032mm (0.008in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals

Formula: 195.08; Form: approx. 1.09g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 010287.G1
also known as 010287-G1
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
1 m
Request bulk or custom format
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
Add to cart
Platinum wire, 0.2032mm (0.008in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number010287.G1
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white wire
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only