Platinum wire, 0.0762mm (0.003in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Platinum wire, 0.0762mm (0.003in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum wire, 0.0762mm (0.003in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals

For ISA Type R or S Standard Grade Thermocouple
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 m
50 cm
10 m
Catalog number 010291.G2
also known as 010291-G2
Price (USD)/ Each
90.40
-
Add to cart
Quantity:
2 m
Request bulk or custom format
Price (USD)/ Each
90.40
Add to cart
Platinum wire, 0.0762mm (0.003in) dia, For ISA Type R or S Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number010291.G2
Price (USD)/ Each
90.40
-
Add to cart
Specifications
Chemical Name or MaterialPlatinum wire
Name Note0.0762mm (0.003 in.) dia.
CAS7440-06-4
Recommended StorageAmbient temperatures
EINECS Number231-116-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only