Thermo Scientific Chemicals

Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min, Thermo Scientific Chemicals

Catalog number: 010301.A7
5 kg, Each
Thermo Scientific Chemicals

Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min, Thermo Scientific Chemicals

Catalog number: 010301.A7
5 kg, Each
Quantity
Catalog number: 010301.A7
also known as 010301-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-28-0
IUPAC Name
phosphoric acid diamine
Molecular Formula
H9N2O4P
InChI Key
MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES
N.N.OP(O)(O)=O
Impurity content
Sodium, Na: ≤0.005% (≤50ppm)
Impurity content
Sulfate, SO₄: ≤0.01% (≤100ppm)
Insoluble material
≤0.005% (≤50ppm)
pH
5% solution, @ 25°C: pH7.7-8.1
Appearance (Color)
White

Description

Ammonium hydrogen phosphate is used as a fertilizer, as a fire retardant, as a yeast nutrient in winemaking and brewing mead, as an additive in some brands of cigarettes, in purifying sugar and to control precipitation of alkali-soluble and acid-insoluble colloidal dyes on wool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text