Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 010308.FF
also known as 010308-FF
Price (USD)/ Each
243.00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
243.00
Add to cart
Rhenium foil, 0.1mm (0.004in) thick, 99.97% (metals basis), Thermo Scientific Chemicals
Catalog number010308.FF
Price (USD)/ Each
243.00
-
Add to cart
Chemical Identifiers
CAS7440-15-5
IUPAC Namerhenium
Molecular FormulaRe
InChI KeyWUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES[Re]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.1 mm (0.004 in.) thick
Total Metal Impurities≤0.03%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only