Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals
Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
10 cm
250 cm
Catalog number 010311.BW
also known as 010311-BW
Price (USD)/ Each
241.00
-
Add to cart
Quantity:
50 cm
Request bulk or custom format
Price (USD)/ Each
241.00
Add to cart
Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals
Catalog number010311.BW
Price (USD)/ Each
241.00
-
Add to cart
Specifications
Chemical Name or MaterialRhenium wire
Name Note0.25mm (0.01 in.) dia.
CAS7440-15-5
Recommended StorageAmbient temperatures
EINECS Number231-124-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only