Rhenium powder, 99.99%, Thermo Scientific Chemicals
Rhenium powder, 99.99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium powder, 99.99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.2 g
10 g
50 g
Catalog number 010313.04
also known as 010313-04
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
Add to cart
Rhenium powder, 99.99%, Thermo Scientific Chemicals
Catalog number010313.04
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-15-5
IUPAC Namerhenium
Molecular FormulaRe
InChI KeyWUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES[Re]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentGray
Total Metal Impurities≤ 0.01%
FormPowder
Particle Size-325 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only